missing translation for 'onlineSavingsMsg'
Learn More

N-Methylbis(trifluoroacetamide) 97.0+%, TCI America™

Catalog No. M06711ML
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M06711ML 1 mL
M06715ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M06711ML Supplier TCI America Supplier No. M06711ML

Chemical Identifiers

CAS 685-27-8
Molecular Formula C5H3F6NO2
Molecular Weight (g/mol) 223.074
MDL Number MFCD00000412
InChI Key AWGBWLXGUPTXHF-UHFFFAOYSA-N
Synonym mbtfa, n-methyl-bis trifluoroacetamide, n-methylbis trifluoroacetamide, 2,2,2-trifluoro-n-methyl-n-trifluoroacetyl acetamide, n,n-bis trifluoroacetyl methylamine, 2,2,2-trifluoro-n-methyl-n-2,2,2-trifluoroacetyl acetamide, acetamide, 2,2,2-trifluoro-n-methyl-n-trifluoroacetyl, n-methylbistrifluoroacetamide, n-methyl-bis-trifluoroacetamide
PubChem CID 69635
IUPAC Name 2,2,2-trifluoro-N-methyl-N-(2,2,2-trifluoroacetyl)acetamide
SMILES CN(C(=O)C(F)(F)F)C(=O)C(F)(F)F

Specifications

Color Yellow
Quantity 1 mL
UN Number 2920
Formula Weight 223.07
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material N-Methylbis(trifluoroacetamide) [Trifluoroacylating Agent]