missing translation for 'onlineSavingsMsg'
Learn More

N-Methylbenzenesulfonamide 98.0+%, TCI America™

Catalog No. M0157500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M0157500G 500 g
M015725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M0157500G Supplier TCI America Supplier No. M0157500G
Only null left

Chemical Identifiers

CAS 5183-78-8
Molecular Formula C7H9NO2S
Molecular Weight (g/mol) 171.21
MDL Number MFCD00025023
InChI Key SVDVKEBISAOWJT-UHFFFAOYSA-N
Synonym benzenesulfonamide, n-methyl, n-methylbenzenesulphonamide, n-phenylsulfonyl methanamine, methyl phenylsulfonyl amine, n-methylphenylsulfonamide, acmc-1avn9, n-methylbenzene sulfonamide, n-methyl benzenesulphonamide, benzenesulfonamide,n-methyl, benzenesulfonyl-n-methylamide
PubChem CID 78858
IUPAC Name N-methylbenzenesulfonamide
SMILES CNS(=O)(=O)C1=CC=CC=C1

Specifications

Color White-Yellow
Quantity 500 g
Formula Weight 171.21
Percent Purity ≥98.0% (GC)
Physical Form Crystals
Chemical Name or Material N-Methylbenzenesulfonamide
EINECSNumber (3)-1930
TSCA Yes