missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Methylbenzenesulfonamide 98.0+%, TCI America™
Specifications
Specifications
| Color | White-Yellow |
| Quantity | 25 g |
| Formula Weight | 171.21 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystals |
| Chemical Name or Material | N-Methylbenzenesulfonamide |
Safety and Handling
| EINECSNumber | (3)-1930 |
| TSCA | Yes |