missing translation for 'onlineSavingsMsg'
Learn More

N-Methylanthranilic acid, 90+%

Catalog No. aab2053136
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2053136 500 g
AAB2053114 25 g
AAB2053122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2053136 Supplier Thermo Scientific Chemicals Supplier No. B2053136
Only null left

CAS: 119-68-6 | C8H9NO2 | 151.165 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 119-68-6
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.165
MDL Number MFCD00002424
InChI Key WVMBPWMAQDVZCM-UHFFFAOYSA-N
Synonym n-methylanthranilic acid, 2-methylamino benzoic acid, n-methyl-2-aminobenzoic acid, n-methyl-o-aminobenzoic acid, benzoic acid, 2-methylamino, o-methylamino benzoic acid, anthranilic acid, n-methyl, 2-methylamino-benzoic acid, kyselina n-methylanthranilova, unii-vpb2514iuj
PubChem CID 67069
ChEBI CHEBI:16394
IUPAC Name 2-(methylamino)benzoic acid
SMILES CNC1=CC=CC=C1C(=O)O

Specifications

Melting Point ∼171°C (decomposition)
Quantity 500 g
Beilstein 2208135
Formula Weight 151.17
Percent Purity ≥90%
Chemical Name or Material N-Methylanthranilic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 204-343-9
RTECSNumber CB3300000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only