missing translation for 'onlineSavingsMsg'
Learn More

N-Methylaniline 98.0+%, TCI America™

Catalog No. p-7132017
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M014725G 25 g
M0147500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M014725G Supplier TCI America Supplier No. M014725G
Only null left

Chemical Identifiers

CAS 100-61-8
Molecular Formula C7H9N
Molecular Weight (g/mol) 107.16
MDL Number MFCD00008283
InChI Key AFBPFSWMIHJQDM-UHFFFAOYSA-N
Synonym methylaniline, monomethylaniline, n-methyl aniline, benzenamine, n-methyl, methylphenylamine, n-methylbenzenamine, n-monomethylaniline, n-methylaminobenzene, methylamino benzene, anilinomethane
PubChem CID 7515
ChEBI CHEBI:15733
IUPAC Name N-methylaniline
SMILES CNC1=CC=CC=C1

Specifications

Melting Point -57°C
Color Yellow
Boiling Point 196°C
Quantity 25 g
UN Number 2294
Formula Weight 107.16
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N-Methylaniline
EINECSNumber (3)-0106
RTECSNumber BY4550000
TSCA Yes