missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Methyl-p-toluenesulfonamide 98.0+%, TCI America™
$50.33 - $301.98
Chemical Identifiers
| CAS | 640-61-9 |
|---|---|
| Molecular Formula | C8H11NO2S |
| Molecular Weight (g/mol) | 185.241 |
| InChI Key | GWLOGZRVYXAHRE-UHFFFAOYSA-N |
| Synonym | n-methyl-p-toluenesulfonamide, benzenesulfonamide, n,4-dimethyl, p-tolylsulfonyl methylamine, p-toluene-n-methylsulfonamide, p-toluenesulfonamide, n-methyl, n,4-dimethyl-benzenesulfonamide, n-methyl-para-toluenesulfonamide, n,4-dimethylbenzene-1-sulfonamide, benzenesulfonamide,4-dimethyl, methyl 4-methylphenyl sulfonyl amine |
| PubChem CID | 12543 |
| IUPAC Name | N,4-dimethylbenzenesulfonamide |
| SMILES | CC1=CC=C(C=C1)S(=O)(=O)NC |
Chemical Identifiers
| 640-61-9 | |
| 185.241 | |
| n-methyl-p-toluenesulfonamide, benzenesulfonamide, n,4-dimethyl, p-tolylsulfonyl methylamine, p-toluene-n-methylsulfonamide, p-toluenesulfonamide, n-methyl, n,4-dimethyl-benzenesulfonamide, n-methyl-para-toluenesulfonamide, n,4-dimethylbenzene-1-sulfonamide, benzenesulfonamide,4-dimethyl, methyl 4-methylphenyl sulfonyl amine | |
| N,4-dimethylbenzenesulfonamide |
| C8H11NO2S | |
| GWLOGZRVYXAHRE-UHFFFAOYSA-N | |
| 12543 | |
| CC1=CC=C(C=C1)S(=O)(=O)NC |
Specifications
| 640-61-9 | |
| White-Yellow | |
| 25 g | |
| GWLOGZRVYXAHRE-UHFFFAOYSA-N | |
| N,4-dimethylbenzenesulfonamide | |
| 12543 | |
| ≥98.0% (HPLC,N) | |
| N-Methyl-p-toluenesulfonamide |
| 79°C | |
| C8H11NO2S | |
| n-methyl-p-toluenesulfonamide, benzenesulfonamide, n,4-dimethyl, p-tolylsulfonyl methylamine, p-toluene-n-methylsulfonamide, p-toluenesulfonamide, n-methyl, n,4-dimethyl-benzenesulfonamide, n-methyl-para-toluenesulfonamide, n,4-dimethylbenzene-1-sulfonamide, benzenesulfonamide,4-dimethyl, methyl 4-methylphenyl sulfonyl amine | |
| CC1=CC=C(C=C1)S(=O)(=O)NC | |
| 185.241 | |
| 185.24 | |
| Crystalline Powder |
Safety and Handling
TSCA : Yes