missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Methyl-N-trimethylsilyltrifluoroacetamide 95.0+%, TCI America™
Supplier: TCI America M067225ML
Specifications
| N-Methyl-N-trimethylsilyltrifluoroacetamide [Trimethylsilylating Agent] | |
| Yellow | |
| C6H12F3NOSi | |
| 25 mL | |
| mstfa, n-methyl-n-trimethylsilyl trifluoroacetamide, 2,2,2-trifluoro-n-methyl-n-trimethylsilyl acetamide, n-methyl-n-trimethylsilyltrifluoroacetamide, acetamide, 2,2,2-trifluoro-n-methyl-n-trimethylsilyl, n-methyl-n-trimethylsilyl-trifluoroacetamide, acmc-209tfm | |
| CN(C(=O)C(F)(F)F)[Si](C)(C)C | |
| 199.25 | |
| CHEBI:85064 | |
| ≥95.0% (GC) |
| 24589-78-4 | |
| 132°C | |
| MFCD00000411 | |
| 1993 | |
| MSPCIZMDDUQPGJ-UHFFFAOYSA-N | |
| 2,2,2-trifluoro-N-methyl-N-(trimethylsilyl)acetamide | |
| 32510 | |
| 199.25 | |
| Liquid |
Chemical Identifiers
| 24589-78-4 | |
| 199.25 | |
| MSPCIZMDDUQPGJ-UHFFFAOYSA-N | |
| 32510 | |
| 2,2,2-trifluoro-N-methyl-N-(trimethylsilyl)acetamide |
| C6H12F3NOSi | |
| MFCD00000411 | |
| mstfa, n-methyl-n-trimethylsilyl trifluoroacetamide, 2,2,2-trifluoro-n-methyl-n-trimethylsilyl acetamide, n-methyl-n-trimethylsilyltrifluoroacetamide, acetamide, 2,2,2-trifluoro-n-methyl-n-trimethylsilyl, n-methyl-n-trimethylsilyl-trifluoroacetamide, acmc-209tfm | |
| CHEBI:85064 | |
| CN(C(=O)C(F)(F)F)[Si](C)(C)C |
Safety and Handling
RTECSNumber : AC9800000
TSCA : Yes