missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-N-trimethylsilyltrifluoroacetamide 95.0+%, TCI America™

Catalog No. p-7132293
Change view
Click to view available options
Quantity:
25 mL
5 mL
Catalog No. Quantity
M06725ML 5 mL
M067225ML 25 mL
2 options

Catalog No. M06725ML

Supplier: TCI America M06725ML

Only null left

Chemical Identifiers

CAS 24589-78-4
Molecular Formula C6H12F3NOSi
Molecular Weight (g/mol) 199.25
MDL Number MFCD00000411
InChI Key MSPCIZMDDUQPGJ-UHFFFAOYSA-N
Synonym mstfa, n-methyl-n-trimethylsilyl trifluoroacetamide, 2,2,2-trifluoro-n-methyl-n-trimethylsilyl acetamide, n-methyl-n-trimethylsilyltrifluoroacetamide, acetamide, 2,2,2-trifluoro-n-methyl-n-trimethylsilyl, n-methyl-n-trimethylsilyl-trifluoroacetamide, acmc-209tfm
PubChem CID 32510
ChEBI CHEBI:85064
IUPAC Name 2,2,2-trifluoro-N-methyl-N-(trimethylsilyl)acetamide
SMILES CN(C(=O)C(F)(F)F)[Si](C)(C)C

Specifications

Color Yellow
Boiling Point 132°C
Quantity 5 mL
UN Number 1993
Formula Weight 199.25
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material N-Methyl-N-trimethylsilyltrifluoroacetamide [Trimethylsilylating Agent]
RTECSNumber AC9800000
TSCA Yes