missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-N-(trimethylsilyl)trifluoroacetamide, 97%

CAS: 24589-78-4 | C6H12F3NOSi | 199.25 g/mol

Supplier:  Thermo Scientific Chemicals 221580250

 View more versions of this product

Catalog No. AC221580250


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Alkylation reagent, Silylation reagent
Specifications

Specifications

N-Methyl-N-(trimethylsilyl)trifluoroacetamide
96.0
Colorless to Yellow
130.0°C to 132.0°C
Authentic
Glass Bottle
1.3782 to 1.3802
MFCD00000411
2920
mstfa, n-methyl-n-trimethylsilyl trifluoroacetamide, 2,2,2-trifluoro-n-methyl-n-trimethylsilyl acetamide, n-methyl-n-trimethylsilyltrifluoroacetamide, acetamide, 2,2,2-trifluoro-n-methyl-n-trimethylsilyl, n-methyl-n-trimethylsilyl-trifluoroacetamide, acmc-209tfm
MSPCIZMDDUQPGJ-UHFFFAOYSA-N
2,2,2-trifluoro-N-methyl-N-trimethylsilylacetamide
32510
199.25
Liquid
24589-78-4
100.0
1.0700g/mL
25°C
96% min. (GC)
C6H12F3NOSi
CF3CON(CH3)Si(CH3)3
25 g
1.07
Solubility in water: reacts
CN(C(=O)C(F)(F)F)[Si](C)(C)C
199.25
CHEBI:85064
97%

Chemical Identifiers

24589-78-4
199.25
MSPCIZMDDUQPGJ-UHFFFAOYSA-N
32510
2,2,2-trifluoro-N-methyl-N-trimethylsilylacetamide
C6H12F3NOSi
MFCD00000411
mstfa, n-methyl-n-trimethylsilyl trifluoroacetamide, 2,2,2-trifluoro-n-methyl-n-trimethylsilyl acetamide, n-methyl-n-trimethylsilyltrifluoroacetamide, acetamide, 2,2,2-trifluoro-n-methyl-n-trimethylsilyl, n-methyl-n-trimethylsilyl-trifluoroacetamide, acmc-209tfm
CHEBI:85064
CN(C(=O)C(F)(F)F)[Si](C)(C)C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapor.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present

GHS Signal Word: Warning

EINECSNumber : 246-331-6

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only