missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-N-phenyl-beta-alanine ethyl ester, 99%, Thermo Scientific Chemicals

Catalog No. AAH6161514
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6161514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6161514 Supplier Thermo Scientific Chemicals Supplier No. H6161514
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2003-76-1 | C12H17NO2 | 207.273 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2003-76-1
Molecular Formula C12H17NO2
Molecular Weight (g/mol) 207.273
MDL Number MFCD01249502
InChI Key KIGKCEWLNGHFCS-UHFFFAOYSA-N
Synonym n-methyl-n-phenyl-beta-alanine ethyl ester, n-methyl-n-2-ethoxycarbonyl ethyl aniline, ethyl 3-methyl phenyl amino propanoate, ethyl 3-n-methylanilino propanoate
PubChem CID 3803593
IUPAC Name ethyl 3-(N-methylanilino)propanoate
SMILES CCOC(=O)CCN(C)C1=CC=CC=C1

Specifications

Boiling Point 98°C to 100°C (0.05mmHg)
Color Orange
Beilstein 2727240
Quantity 25 g
Formula Weight 207.27
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material N-Methyl-N-phenyl-beta-alanine ethyl ester
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only