accessibility menu, dialog, popup

UserName

H3COONCH3

CAS RN 2003-76-1

IUPAC Name: ethyl 3-[methyl(phenyl)amino]propanoate
Synonyms: ethyl 3-[methyl(phenyl)amino]propanoate
Molecular Weight (g/mol): 207.27
Molecular Formula: C12H17NO2
InChi Key: KIGKCEWLNGHFCS-UHFFFAOYSA-N
SMILES: CCOC(=O)CCN(C)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)