missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-N-{[5-(pyridin-3-yloxy)-2-furyl] methyl}amine, 97%, Thermo Scientific™

Catalog No. CC42746CB
Change view
Click to view available options
Quantity:
250mg
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC42746CB 250mg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC42746CB Supplier Thermo Scientific Chemicals Supplier No. CC42746CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 859850-77-4
Molecular Formula C11H12N2O2
Molecular Weight (g/mol) 204.229
MDL Number MFCD07772853
InChI Key MUKCVLFJCGZPKF-UHFFFAOYSA-N
Synonym n-methyl-n-5-pyridin-3-yloxy-2-furyl methyl amine, methyl 5-pyridin-3-yloxy furan-2-yl methyl amine, n-methyl-1-5-pyridin-3-yloxy furan-2-yl methanamine, methyl 5-3-pyridyloxy 2-furyl methyl amine, 2-furanmethanamine,n-methyl-5-3-pyridinyloxy, n-methyl-1-5-pyridin-3-yloxyfuran-2-yl methanamine, n-methyl-1-5-pyridin-3-yl oxy furan-2-yl methanamine
PubChem CID 7164635
IUPAC Name N-methyl-1-(5-pyridin-3-yloxyfuran-2-yl)methanamine
SMILES CNCC1=CC=C(O1)OC2=CN=CC=C2

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250mg
Formula Weight 204.23
Percent Purity 97%
Chemical Name or Material N-methyl-N-{[5-(pyridin-3-yloxy)-2-furyl]methyl}amine
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if inhaled.
Harmful in contact with skin.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.