accessibility menu, dialog, popup

UserName

CH3NHOON

CAS RN 859850-77-4

IUPAC Name: methyl({[5-(pyridin-3-yloxy)furan-2-yl]methyl})amine
Synonyms: methyl({[5-(pyridin-3-yloxy)furan-2-yl]methyl})amine
Molecular Weight (g/mol): 204.23
Molecular Formula: C11H12N2O2
InChi Key: MUKCVLFJCGZPKF-UHFFFAOYSA-N
SMILES: CNCC1=CC=C(OC2=CN=CC=C2)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)