missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-N-(4-pyridin-4-ylbenzyl)amine, 90%, Thermo Scientific™

Catalog No. p-4510546
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC35446CB 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC35446CB Supplier Thermo Scientific Chemicals Supplier No. CC35446CB

Chemical Identifiers

CAS 852180-64-4
Molecular Formula C13H14N2
Molecular Weight (g/mol) 198.269
InChI Key MDKJAMMSIZLRBH-UHFFFAOYSA-N
Synonym n-methyl-n-4-pyridin-4-ylbenzyl amine, n-methyl-4-pyridin-4-yl benzylamine, methyl 4-pyridin-4-yl phenyl methyl amine, n-methyl-1-4-pyridin-4-yl phenyl methanamine, n-methyl-1-4-pyridin-4-ylphenyl methanamine, methyl 4-4-pyridyl phenyl methyl amine, benzenemethanamine,n-methyl-4-4-pyridinyl
PubChem CID 7060572
IUPAC Name N-methyl-1-(4-pyridin-4-ylphenyl)methanamine
SMILES CNCC1=CC=C(C=C1)C2=CC=NC=C2

Specifications

Assay Percent Range 90%
Quantity 250 mg
Formula Weight 198.27
Percent Purity 90%
Chemical Name or Material N-Methyl-N-(4-pyridin-4-ylbenzyl)amine