accessibility menu, dialog, popup

UserName

CH3HNN

CAS RN 852180-64-4

IUPAC Name: methyl({[4-(pyridin-4-yl)phenyl]methyl})amine
Synonyms: methyl({[4-(pyridin-4-yl)phenyl]methyl})amine
Molecular Weight (g/mol): 198.27
Molecular Formula: C13H14N2
InChi Key: MDKJAMMSIZLRBH-UHFFFAOYSA-N
SMILES: CNCC1=CC=C(C=C1)C1=CC=NC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results