missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-N-[4-(2-morpholin-4-ylethoxy)benzyl]amine, 95%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC42146DE

 View more versions of this product

Catalog No. CC42146DE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

N-Methyl-N-[4-(2-morpholin-4-ylethoxy)benzyl]amine
94.5
95%
MFCD07368554
n-methyl-n-4-2-morpholin-4-ylethoxy benzyl amine, methyl 4-2-morpholin-4-yl ethoxy phenyl methyl amine, benzenemethanamine,n-methyl-4-2-4-morpholinyl ethoxy, n-methyl-1-4-2-morpholin-4-ylethoxy phenyl methanamine, n-methyl-n-4-2-morpholin-4-ylethoxy benzylamine, n-methyl-1-4-2-morpholinoethoxy phenyl methanamine, methyl 4-2-morpholin-4-ylethoxy phenyl methyl amine
CNCC1=CC=C(C=C1)OCCN2CCOCC2
250.342
250.34
852180-77-9
100.0
C14H22N2O2
5g
CFDWJPMGCABITD-UHFFFAOYSA-N
N-methyl-1-[4-(2-morpholin-4-ylethoxy)phenyl]methanamine
7060587
95%

Chemical Identifiers

852180-77-9
250.342
CFDWJPMGCABITD-UHFFFAOYSA-N
7060587
CNCC1=CC=C(C=C1)OCCN2CCOCC2
C14H22N2O2
MFCD07368554
n-methyl-n-4-2-morpholin-4-ylethoxy benzyl amine, methyl 4-2-morpholin-4-yl ethoxy phenyl methyl amine, benzenemethanamine,n-methyl-4-2-4-morpholinyl ethoxy, n-methyl-1-4-2-morpholin-4-ylethoxy phenyl methanamine, n-methyl-n-4-2-morpholin-4-ylethoxy benzylamine, n-methyl-1-4-2-morpholinoethoxy phenyl methanamine, methyl 4-2-morpholin-4-ylethoxy phenyl methyl amine
N-methyl-1-[4-(2-morpholin-4-ylethoxy)phenyl]methanamine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS Documents

Documents

Promotions

Promotions