missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-N-(2-pyridyl)formamide 98.0+%, TCI America™

Catalog No. M11875G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M11875G 5 g
M11871G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M11875G Supplier TCI America Supplier No. M11875G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 67242-59-5
Molecular Formula C7H8N2O
Molecular Weight (g/mol) 136.154
MDL Number MFCD00006260
InChI Key DIRVEOCOAHGVJV-UHFFFAOYSA-N
PubChem CID 572611
IUPAC Name N-methyl-N-pyridin-2-ylformamide
SMILES CN(C=O)C1=CC=CC=N1

Specifications

Color Yellow
Boiling Point 82°C
Quantity 5 g
Synonym 2-(N-Formyl-N-methylamino)pyridine, Meyer′s Reagent
Formula Weight 136.15
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material N-Methyl-N-(2-pyridyl)formamide