CAS RN 67242-59-5
CAS RN 67242-59-5
IUPAC Name:
N-methyl-N-(pyridin-2-yl)formamide
Synonyms:
N-methyl-N-(pyridin-2-yl)formamide
Molecular Weight (g/mol):
136.15
Molecular Formula:
C7H8N2O
InChi Key:
DIRVEOCOAHGVJV-UHFFFAOYSA-N
SMILES:
CN(C=O)C1=CC=CC=N1
1
–
1
of
1
results
1
N-Methyl-N-(2-pyridyl)formamide 98.0+%, TCI America™
CAS: 67242-59-5 Molecular Formula: C7H8N2O Molecular Weight (g/mol): 136.154 MDL Number: MFCD00006260 InChI Key: DIRVEOCOAHGVJV-UHFFFAOYSA-N PubChem CID: 572611 IUPAC Name: N-methyl-N-pyridin-2-ylformamide SMILES: CN(C=O)C1=CC=CC=N1