missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-N'-(2,4-xylyl)formamidine, MilliporeSigma™ Supelco™

Catalog No. 111002161
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111002161 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111002161 Supplier Merck Emd Millipore Supplier No. BP641
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Specifications

CAS 33089-74-6
Linear Formula C10H14N2
MDL Number MFCD00225755
Synonym Amitraz Metabolite BTS 27271; N-(2,4-Dimethylphenyl)-N′-methylformamidine; N-Methyl-N′-2,4-xylylformamidine
Type Pharmacopeia and Metrological Institutes Standards
Quantity 25 mg
Formula Weight 162.23
Grade Pharmaceutical Primary Standard