missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Methyl-m-toluidine 98.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 207°C |
| Quantity | 500 mL |
| UN Number | 2810 |
| Formula Weight | 121.18 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Liquid |
| Chemical Name or Material | N-Methyl-m-toluidine |
Safety and Handling
| EINECSNumber | (3)-0129&(3)-0187 |
| TSCA | Yes |