missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-epsilon-caprolactam 97.0+%, TCI America™

Catalog No. M068625G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M068625G 25 g
M0686500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M068625G Supplier TCI America Supplier No. M068625G

Chemical Identifiers

CAS 2556-73-2
Molecular Formula C7H13NO
Molecular Weight (g/mol) 127.187
MDL Number MFCD00003263
InChI Key ZWXPDGCFMMFNRW-UHFFFAOYSA-N
Synonym n-methylcaprolactam, 1-methylcaprolactam, n-methyl-.epsilon.-caprolactam, 2h-azepin-2-one, hexahydro-1-methyl, n-methyl-e-caprolactam, epsilon-caprolactam, n-methyl, 2h-azepin-2-one, hexahydro-n-methyl, hexahydro-n-methyl-2h-azepin-2-one, n-methyl-epsilon-caprolactam, n-methyl caprolactam
PubChem CID 17369
IUPAC Name 1-methylazepan-2-one
SMILES CN1CCCCCC1=O

Specifications

Color Yellow
Boiling Point 120°C
Quantity 25 g
Formula Weight 127.19
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material N-Methyl-epsilon-caprolactam
RTECSNumber CM4025000
TSCA No