missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl(diphenylmethyl)amine, 98%

Catalog No. p-7033252
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5032703 1 g
AAH5032706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5032703 Supplier Thermo Scientific Chemicals Supplier No. H5032703

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 14683-47-7 | C14H15N | 197.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14683-47-7
Molecular Formula C14H15N
Molecular Weight (g/mol) 197.28
MDL Number MFCD00467853
InChI Key SHDMMLFAFLZUEV-UHFFFAOYSA-N
Synonym n-diphenylmethyl methylamine, methylbenzhydrylamine, n-benzhydryl-n-methylamine, n-methylbenzhydrylamine, diphenylmethyl methyl amine, diphenyl-methyl methyl amine, diphenylmethyl methylamine, n-methyldiphenylmethanamine, acmc-20akaj, n-methyl benzhydrylamine
PubChem CID 411467
IUPAC Name N-methyl-1,1-diphenylmethanamine
SMILES CNC(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 38°C to 42°C
Quantity 1 g
Beilstein 2096409
Formula Weight 197.28
Percent Purity 98%
Chemical Name or Material N-Methyl(diphenylmethyl)amine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only