missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Methyl-D-glucamine 99.0+%, TCI America™
Specifications
Specifications
| Melting Point | 131°C |
| Color | White |
| Quantity | 500 g |
| Formula Weight | 195.22 |
| Percent Purity | ≥99.0% (T) |
| Grade | Not Available |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | N-Methyl-D-glucamine |
Safety and Handling
| EINECSNumber | (2)-2412 |
| TSCA | Yes |