missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-D-glucamine 99.0+%, TCI America™

Catalog No. M022725G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M022725G 25 g
M0227500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M022725G Supplier TCI America Supplier No. M022725G

Chemical Identifiers

CAS 6284-40-8
Molecular Formula C7H17NO5
Molecular Weight (g/mol) 195.215
MDL Number MFCD00004707
InChI Key MBBZMMPHUWSWHV-BDVNFPICSA-N
Synonym meglumine, n-methyl-d-glucamine, n-methylglucamine, meglumin, 1-deoxy-1-methylamino-d-glucitol, methylglucamin, meglumina, 1-deoxy-1-methylaminosorbitol, d-glucitol, 1-deoxy-1-methylamino, n-methyl-d--glucamine
PubChem CID 8567
ChEBI CHEBI:59732
IUPAC Name (2R,3R,4R,5S)-6-(methylamino)hexane-1,2,3,4,5-pentol
SMILES CNCC(C(C(C(CO)O)O)O)O

Specifications

Melting Point 131°C
Color White
Quantity 25 g
Formula Weight 195.22
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Methyl-D-glucamine
EINECSNumber (2)-2412
TSCA Yes