Learn More
Thermo Scientific Chemicals N-Methyl-D-glucamine, 99%
N-Methyl-D-glucamine, CAS # 6284-40-8, also known as meglumine, is a base compound commonly used in combination with iodinated organic compounds in contrast medium. | CAS: 6284-40-8 | C7H17NO5 | 195.21 g/mol
Supplier: Thermo Scientific Chemicals 126845000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Information
• N-Methyl-D-glucamine is a hexosamine compound that is frequently used in combination with iodinated organic compounds in contrast media such as iothalamate meglumine, iodipamide meglumine, and diatrizoate meglumine
Applications
• It is also useful when applied in ion-exchange resins to chelate boron ions selectively
• It is considered a base and frequently used in the preparation of salts of certain acids
• It is also applied in the synthesis of surface-active agents and laboratory dyes
Specifications
| N-Methyl-D-glucamine | |
| 6284-40-8 | |
| 0.03 max. (20 w/v% aq. soln.) at 420nm | |
| 128.0°C to 132.0°C | |
| 1% max. (1 g, 100°C, 3 hrs) | |
| 99% | |
| C7H17NO5 | |
| 500 g | |
| 15, 6150 | |
| − 16.50 | |
| Solubility in water: 100 g/100 mL water (25°C). Other solubilities: 1.2 g/100 mL alcohol (25°C), 21 g/100 mL alcohol (70°C) | |
| CNCC(C(C(C(CO)O)O)O)O | |
| 195.21 | |
| CHEBI:59732 | |
| 99% |
| 99% | |
| 98.5 | |
| 100.0 | |
| White | |
| Authentic | |
| Plastic bottle | |
| MFCD00004707 | |
| 04, IV, 1914 | |
| − 16.50 (20.00°C c=10,H2O,on dry sub) | |
| meglumine, n-methyl-d-glucamine, n-methylglucamine, meglumin, 1-deoxy-1-methylamino-d-glucitol, methylglucamin, meglumina, 1-deoxy-1-methylaminosorbitol, d-glucitol, 1-deoxy-1-methylamino, n-methyl-d--glucamine | |
| MBBZMMPHUWSWHV-BDVNFPICSA-N | |
| (2R,3R,4R,5S)-6-(methylamino)hexane-1,2,3,4,5-pentol | |
| 8567 | |
| 195.21 | |
| Crystalline Powder |
Chemical Identifiers
| 6284-40-8 | |
| 195.21 | |
| MBBZMMPHUWSWHV-BDVNFPICSA-N | |
| 8567 | |
| (2R,3R,4R,5S)-6-(methylamino)hexane-1,2,3,4,5-pentol |
| C7H17NO5 | |
| MFCD00004707 | |
| meglumine, n-methyl-d-glucamine, n-methylglucamine, meglumin, 1-deoxy-1-methylamino-d-glucitol, methylglucamin, meglumina, 1-deoxy-1-methylaminosorbitol, d-glucitol, 1-deoxy-1-methylamino, n-methyl-d--glucamine | |
| CHEBI:59732 | |
| CNCC(C(C(C(CO)O)O)O)O |
Safety and Handling
AIR SENSITIVE
HYGROSCOPIC
EINECSNumber : 228-506-9
TSCA : TSCA
RUO – Research Use Only