missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Methyl-D-aspartic acid, 98+%
CAS: 6384-92-5 | C5H9NO4 | 147.13 g/mol
Supplier: Thermo Scientific Chemicals J61361MC
| Quantity | 100 mg |
|---|
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| C5H9NO4 | |
| MFCD00004226 | |
| n-methyl-d-aspartic acid, nmda, n-methyl-d-aspartate, n-methylaspartate, d-aspartic acid, n-methyl, methyl aspartic acid, n-me-d-asp-oh, r-2-methylamino succinic acid, unii-1903b9q6pi, n methyl d aspartate | |
| CHEBI:31882 | |
| CNC(CC(=O)O)C(=O)O |
Specifications
| 187°C to 192°C | |
| 6384-92-5 | |
| C5H9NO4 | |
| 1724431 | |
| n-methyl-d-aspartic acid, nmda, n-methyl-d-aspartate, n-methylaspartate, d-aspartic acid, n-methyl, methyl aspartic acid, n-me-d-asp-oh, r-2-methylamino succinic acid, unii-1903b9q6pi, n methyl d aspartate | |
| CNC(CC(=O)O)C(=O)O | |
| 100 mg | |
| 22880 | |
| 147.13 | |
| ≥98% | |
| Powder |
| N-Methyl-D-aspartic acid | |
| White | |
| MFCD00004226 | |
| 14,6662 | |
| HOKKHZGPKSLGJE-GSVOUGTGSA-N | |
| (2R)-2-(methylamino)butanedioic acid | |
| 147.13 | |
| CHEBI:31882 | |
| −15° (c=2 in Water) | |
| Hygroscopic |
Safety and Handling
RTECSNumber : CI9457000
TSCA : No
Recommended Storage : Keep cold