missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-(7-bromo-2,3-dihydrothieno[3,4-b][1,4]dioxin-5-yl)methylamine, 97%, Thermo Scientific™

Chemical Identifiers

CAS 886851-54-3
Molecular Formula C8H10BrNO2S
Molecular Weight (g/mol) 264.137
MDL Number MFCD08690308
InChI Key FJHBRYOEMIQSPC-UHFFFAOYSA-N
Synonym n-methyl-7-bromo-2,3-dihydrothieno 3,4-b 1,4 dioxin-5-yl methylamine, n-methyl-5-aminomethyl-7-bromo-2,3-dihydrothieno 3,4-b 1,4 dioxine, 7-bromo-2h,3h-thieno 3,4-b 1,4 dioxin-5-yl methyl methyl amine, 7-bromo 2h,3h-thiopheno 4,3-e 1,4-dioxan-5-yl methyl methylamine, thieno 3,4-b-1,4-dioxin-5-methanamine,7-bromo-2,3-dihydro-n-methyl, 1-7-bromo-2,3-dihydrothieno 3,4-b 1,4 dioxin-5-yl-n-methylmethanamine
PubChem CID 18525892
IUPAC Name 1-(5-bromo-2,3-dihydrothieno[3,4-b][1,4]dioxin-7-yl)-N-methylmethanamine
SMILES CNCC1=C2C(=C(S1)Br)OCCO2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
CC58146CB
View Documents
N/A 250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC58146DA
View Documents
N/A 1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

886851-54-3
264.137
FJHBRYOEMIQSPC-UHFFFAOYSA-N
18525892
CNCC1=C2C(=C(S1)Br)OCCO2
C8H10BrNO2S
MFCD08690308
n-methyl-7-bromo-2,3-dihydrothieno 3,4-b 1,4 dioxin-5-yl methylamine, n-methyl-5-aminomethyl-7-bromo-2,3-dihydrothieno 3,4-b 1,4 dioxine, 7-bromo-2h,3h-thieno 3,4-b 1,4 dioxin-5-yl methyl methyl amine, 7-bromo 2h,3h-thiopheno 4,3-e 1,4-dioxan-5-yl methyl methylamine, thieno 3,4-b-1,4-dioxin-5-methanamine,7-bromo-2,3-dihydro-n-methyl, 1-7-bromo-2,3-dihydrothieno 3,4-b 1,4 dioxin-5-yl-n-methylmethanamine
1-(5-bromo-2,3-dihydrothieno[3,4-b][1,4]dioxin-7-yl)-N-methylmethanamine

Specifications

886851-54-3
100.0
C8H10BrNO2S
n-methyl-7-bromo-2,3-dihydrothieno 3,4-b 1,4 dioxin-5-yl methylamine, n-methyl-5-aminomethyl-7-bromo-2,3-dihydrothieno 3,4-b 1,4 dioxine, 7-bromo-2h,3h-thieno 3,4-b 1,4 dioxin-5-yl methyl methyl amine, 7-bromo 2h,3h-thiopheno 4,3-e 1,4-dioxan-5-yl methyl methylamine, thieno 3,4-b-1,4-dioxin-5-methanamine,7-bromo-2,3-dihydro-n-methyl, 1-7-bromo-2,3-dihydrothieno 3,4-b 1,4 dioxin-5-yl-n-methylmethanamine
CNCC1=C2C(=C(S1)Br)OCCO2
264.137
264.14
N-methyl-(7-bromo-2,3-dihydrothieno[3,4-b][1,4]dioxin-5-yl)methylamine
89.5
90%
MFCD08690308
FJHBRYOEMIQSPC-UHFFFAOYSA-N
1-(5-bromo-2,3-dihydrothieno[3,4-b][1,4]dioxin-7-yl)-N-methylmethanamine
18525892
90%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents