missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-(7-bromo-2,3-dihydrothieno[3,4-b][1,4]dioxin-5-yl)methylamine, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC58146DA

Quantity 1g
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. CC58146DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

886851-54-3
264.137
FJHBRYOEMIQSPC-UHFFFAOYSA-N
18525892
CNCC1=C2C(=C(S1)Br)OCCO2
C8H10BrNO2S
MFCD08690308
n-methyl-7-bromo-2,3-dihydrothieno 3,4-b 1,4 dioxin-5-yl methylamine, n-methyl-5-aminomethyl-7-bromo-2,3-dihydrothieno 3,4-b 1,4 dioxine, 7-bromo-2h,3h-thieno 3,4-b 1,4 dioxin-5-yl methyl methyl amine, 7-bromo 2h,3h-thiopheno 4,3-e 1,4-dioxan-5-yl methyl methylamine, thieno 3,4-b-1,4-dioxin-5-methanamine,7-bromo-2,3-dihydro-n-methyl, 1-7-bromo-2,3-dihydrothieno 3,4-b 1,4 dioxin-5-yl-n-methylmethanamine
1-(5-bromo-2,3-dihydrothieno[3,4-b][1,4]dioxin-7-yl)-N-methylmethanamine

Specifications

N-methyl-(7-bromo-2,3-dihydrothieno[3,4-b][1,4]dioxin-5-yl)methylamine
89.5
90%
C8H10BrNO2S
MFCD08690308
FJHBRYOEMIQSPC-UHFFFAOYSA-N
1-(5-bromo-2,3-dihydrothieno[3,4-b][1,4]dioxin-7-yl)-N-methylmethanamine
18525892
90%
886851-54-3
100.0
Amber Glass Bottle
1g
n-methyl-7-bromo-2,3-dihydrothieno 3,4-b 1,4 dioxin-5-yl methylamine, n-methyl-5-aminomethyl-7-bromo-2,3-dihydrothieno 3,4-b 1,4 dioxine, 7-bromo-2h,3h-thieno 3,4-b 1,4 dioxin-5-yl methyl methyl amine, 7-bromo 2h,3h-thiopheno 4,3-e 1,4-dioxan-5-yl methyl methylamine, thieno 3,4-b-1,4-dioxin-5-methanamine,7-bromo-2,3-dihydro-n-methyl, 1-7-bromo-2,3-dihydrothieno 3,4-b 1,4 dioxin-5-yl-n-methylmethanamine
CNCC1=C2C(=C(S1)Br)OCCO2
264.137
264.14
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents