missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-4-pyrimidin-2-ylbenzylamine, 90%, Thermo Scientific™

Catalog No. p-3541131
Change view
Click to view available options
Quantity:
250 mg
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC56246CB 250 mg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC56246CB Supplier Thermo Scientific Chemicals Supplier No. CC56246CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 886851-48-5
Molecular Formula C12H13N3
Molecular Weight (g/mol) 199.257
MDL Number MFCD09702382
InChI Key DIWVUAYHEYROLA-UHFFFAOYSA-N
Synonym n-methyl-4-pyrimidin-2-ylbenzylamine, n-methyl-1-4-pyrimidin-2-yl phenyl methanamine, methyl 4-pyrimidin-2-yl phenyl methyl amine, n-methyl-4-pyrimidin-2-yl benzylamine, benzenemethanamine,n-methyl-4-2-pyrimidinyl, methyl 4-pyrimidin-2-ylphenyl methyl amine, benzenemethanamine, n-methyl-4-2-pyrimidinyl-9ci
PubChem CID 24229561
IUPAC Name N-methyl-1-(4-pyrimidin-2-ylphenyl)methanamine
SMILES CNCC1=CC=C(C=C1)C2=NC=CC=N2

Specifications

CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 199.26
Percent Purity 90%
Chemical Name or Material N-methyl-4-pyrimidin-2-ylbenzylamine
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.