missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-3-nitroaniline 98.0+%, TCI America™

Catalog No. p-7133552
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
Catalog No. Quantity
M30421G 1 g
M30425G 5 g
2 options

Catalog No. M30421G

Supplier: TCI America M30421G

Only null left

Chemical Identifiers

CAS 619-26-1
Molecular Formula C7H8N2O2
Molecular Weight (g/mol) 152.153
MDL Number MFCD00963641
InChI Key IKSRCCUOUJJGAU-UHFFFAOYSA-N
Synonym 3-nitro-n-methylaniline, n-methyl-m-nitroaniline, benzenamine, n-methyl-3-nitro, n-methyl-3-nitro-aniline, acmc-1b45l, n-methyl-n-3-nitrophenyl amine #
PubChem CID 219622
IUPAC Name N-methyl-3-nitroaniline
SMILES CNC1=CC(=CC=C1)[N+](=O)[O-]

Specifications

Melting Point 67°C
Quantity 1 g
Formula Weight 152.15
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-Methyl-3-nitroaniline