accessibility menu, dialog, popup

UserName

CH3HNNOO

CAS RN 619-26-1

IUPAC Name: N-methyl-3-nitroaniline
Synonyms: N-methyl-3-nitroaniline
Molecular Weight (g/mol): 152.15
Molecular Formula: C7H8N2O2
InChi Key: IKSRCCUOUJJGAU-UHFFFAOYSA-N
SMILES: CNC1=CC=CC(=C1)[N+]([O-])=O
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)