Learn More
N-Methyl-2-(tetrahydropyran-4-yloxy)benzylamine, 95%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC52246DA
Specifications
| N-Methyl-2-(tetrahydropyran-4-yloxy)benzylamine | |
| 94.5 | |
| 95% | |
| C13H19NO2 | |
| 1g | |
| ZTMRIJRRCHAUNA-UHFFFAOYSA-N | |
| N-methyl-1-[2-(oxan-4-yloxy)phenyl]methanamine | |
| 24229518 | |
| 95% |
| 906352-71-4 | |
| 100.0 | |
| Amber Glass Bottle | |
| MFCD09064962 | |
| n-methyl-2-tetrahydropyran-4-yloxy benzylamine, n-methyl-1-2-tetrahydro-2h-pyran-4-yl oxy phenyl methanamine, methyl 2-oxan-4-yloxy phenyl methyl amine, n-methyl-n-2-tetrahydro-2h-pyran-4-yloxy benzyl amine, n-methyl-1-2-oxan-4-yl oxy phenyl methanamine, methyl 2-2h-3,4,5,6-tetrahydropyran-4-yloxy phenyl methyl amine, n-methyl-1-2-tetrahydro-2h-pyran-4-yl ;oxy ;phenyl ;methanamine | |
| CNCC1=CC=CC=C1OC2CCOCC2 | |
| 221.3 | |
| 221.3 |
Chemical Identifiers
| 906352-71-4 | |
| 221.3 | |
| ZTMRIJRRCHAUNA-UHFFFAOYSA-N | |
| 24229518 | |
| CNCC1=CC=CC=C1OC2CCOCC2 |
| C13H19NO2 | |
| MFCD09064962 | |
| n-methyl-2-tetrahydropyran-4-yloxy benzylamine, n-methyl-1-2-tetrahydro-2h-pyran-4-yl oxy phenyl methanamine, methyl 2-oxan-4-yloxy phenyl methyl amine, n-methyl-n-2-tetrahydro-2h-pyran-4-yloxy benzyl amine, n-methyl-1-2-oxan-4-yl oxy phenyl methanamine, methyl 2-2h-3,4,5,6-tetrahydropyran-4-yloxy phenyl methyl amine, n-methyl-1-2-tetrahydro-2h-pyran-4-yl ;oxy ;phenyl ;methanamine | |
| N-methyl-1-[2-(oxan-4-yloxy)phenyl]methanamine |
Safety and Handling
GHS H Statement:
Causes serious eye damage.
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger