missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-(2-morpholinopyrimidin-5-yl)methylamine, 97%, Thermo Scientific™

Chemical Identifiers

CAS 937796-14-0
Molecular Formula C10H16N4O
Molecular Weight (g/mol) 208.265
MDL Number MFCD09966161
InChI Key PFYPMSRZQMFCJO-UHFFFAOYSA-N
Synonym n-methyl-2-morpholinopyrimidin-5-yl methylamine, methyl 2-morpholin-4-yl pyrimidin-5-yl methyl amine, methyl 2-morpholin-4-ylpyrimidin-5-yl methyl amine, n-methyl-1-2-morpholin-4-yl pyrimidin-5-yl methanamine
PubChem CID 42556099
IUPAC Name N-methyl-1-(2-morpholin-4-ylpyrimidin-5-yl)methanamine
SMILES CNCC1=CN=C(N=C1)N2CCOCC2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC69546CB
SDS View Documents
Thermo Scientific Chemicals
CC69546CB
250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC69546DA
SDS View Documents
Thermo Scientific Chemicals
CC69546DA
1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

937796-14-0
208.265
PFYPMSRZQMFCJO-UHFFFAOYSA-N
42556099
CNCC1=CN=C(N=C1)N2CCOCC2
C10H16N4O
MFCD09966161
n-methyl-2-morpholinopyrimidin-5-yl methylamine, methyl 2-morpholin-4-yl pyrimidin-5-yl methyl amine, methyl 2-morpholin-4-ylpyrimidin-5-yl methyl amine, n-methyl-1-2-morpholin-4-yl pyrimidin-5-yl methanamine
N-methyl-1-(2-morpholin-4-ylpyrimidin-5-yl)methanamine

Specifications

937796-14-0
100.0
Amber Glass Bottle
MFCD09966161
n-methyl-2-morpholinopyrimidin-5-yl methylamine, methyl 2-morpholin-4-yl pyrimidin-5-yl methyl amine, methyl 2-morpholin-4-ylpyrimidin-5-yl methyl amine, n-methyl-1-2-morpholin-4-yl pyrimidin-5-yl methanamine
CNCC1=CN=C(N=C1)N2CCOCC2
208.265
208.26
N-methyl-(2-morpholinopyrimidin-5-yl)methylamine
96.5
97%
C10H16N4O
250mg
PFYPMSRZQMFCJO-UHFFFAOYSA-N
N-methyl-1-(2-morpholin-4-ylpyrimidin-5-yl)methanamine
42556099
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
missing translation for 'documents'

missing translation for 'documents'