missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-(2-morpholinopyrimidin-5-yl)methylamine, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC69546DA

 View more versions of this product

Catalog No. CC69546DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

N-methyl-(2-morpholinopyrimidin-5-yl)methylamine
96.5
97%
C10H16N4O
1g
PFYPMSRZQMFCJO-UHFFFAOYSA-N
N-methyl-1-(2-morpholin-4-ylpyrimidin-5-yl)methanamine
42556099
97%
937796-14-0
100.0
Amber Glass Bottle
MFCD09966161
n-methyl-2-morpholinopyrimidin-5-yl methylamine, methyl 2-morpholin-4-yl pyrimidin-5-yl methyl amine, methyl 2-morpholin-4-ylpyrimidin-5-yl methyl amine, n-methyl-1-2-morpholin-4-yl pyrimidin-5-yl methanamine
CNCC1=CN=C(N=C1)N2CCOCC2
208.265
208.26

Chemical Identifiers

937796-14-0
208.265
PFYPMSRZQMFCJO-UHFFFAOYSA-N
42556099
CNCC1=CN=C(N=C1)N2CCOCC2
C10H16N4O
MFCD09966161
n-methyl-2-morpholinopyrimidin-5-yl methylamine, methyl 2-morpholin-4-yl pyrimidin-5-yl methyl amine, methyl 2-morpholin-4-ylpyrimidin-5-yl methyl amine, n-methyl-1-2-morpholin-4-yl pyrimidin-5-yl methanamine
N-methyl-1-(2-morpholin-4-ylpyrimidin-5-yl)methanamine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS Documents

Documents

Promotions

Promotions