Learn More
N-Methyl(2-methyl-3-furyl)methylamine, 95%, Thermo Scientific™
Chemical Identifiers
| CAS | 916766-85-3 |
|---|---|
| Molecular Formula | C7H11NO |
| Molecular Weight (g/mol) | 125.171 |
| MDL Number | MFCD09702377 |
| InChI Key | IDODRBWAEHXURE-UHFFFAOYSA-N |
| Synonym | n-methyl 2-methyl-3-furyl methylamine, methyl 2-methylfuran-3-yl methyl amine, n-methyl-2-methylfur-3-yl methylamine, 2-methyl-3-methylamino methyl furan, 3-furanmethanamine,n,2-dimethyl, 3-furanmethanamine, n,2-dimethyl, methyl 2-methyl 3-furyl methyl amine, n-methyl-1-2-methylfuran-3-yl methanamine |
| PubChem CID | 24229548 |
| IUPAC Name | N-methyl-1-(2-methylfuran-3-yl)methanamine |
| SMILES | CC1=C(C=CO1)CNC |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
CC55446CB
|
N/A | 250mg | Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
CC55446DA
|
N/A | 1g | Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 916766-85-3 | |
| 125.171 | |
| IDODRBWAEHXURE-UHFFFAOYSA-N | |
| 24229548 | |
| CC1=C(C=CO1)CNC |
| C7H11NO | |
| MFCD09702377 | |
| n-methyl 2-methyl-3-furyl methylamine, methyl 2-methylfuran-3-yl methyl amine, n-methyl-2-methylfur-3-yl methylamine, 2-methyl-3-methylamino methyl furan, 3-furanmethanamine,n,2-dimethyl, 3-furanmethanamine, n,2-dimethyl, methyl 2-methyl 3-furyl methyl amine, n-methyl-1-2-methylfuran-3-yl methanamine | |
| N-methyl-1-(2-methylfuran-3-yl)methanamine |
Specifications
| 916766-85-3 | |
| 100.0 | |
| C7H11NO | |
| n-methyl 2-methyl-3-furyl methylamine, methyl 2-methylfuran-3-yl methyl amine, n-methyl-2-methylfur-3-yl methylamine, 2-methyl-3-methylamino methyl furan, 3-furanmethanamine,n,2-dimethyl, 3-furanmethanamine, n,2-dimethyl, methyl 2-methyl 3-furyl methyl amine, n-methyl-1-2-methylfuran-3-yl methanamine | |
| CC1=C(C=CO1)CNC | |
| 125.171 | |
| 125.17 | |
| N-methyl(2-methyl-3-furyl)methylamine |
| 94.5 | |
| 95% | |
| MFCD09702377 | |
| IDODRBWAEHXURE-UHFFFAOYSA-N | |
| N-methyl-1-(2-methylfuran-3-yl)methanamine | |
| 24229548 | |
| 95% |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Harmful if inhaled.
Causes severe skin burns and eye damage.
Harmful in contact with skin.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger