missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl(2-methyl-3-furyl)methylamine, 95%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC55446DA

Quantity 1g
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. CC55446DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

916766-85-3
125.171
IDODRBWAEHXURE-UHFFFAOYSA-N
24229548
CC1=C(C=CO1)CNC
C7H11NO
MFCD09702377
n-methyl 2-methyl-3-furyl methylamine, methyl 2-methylfuran-3-yl methyl amine, n-methyl-2-methylfur-3-yl methylamine, 2-methyl-3-methylamino methyl furan, 3-furanmethanamine,n,2-dimethyl, 3-furanmethanamine, n,2-dimethyl, methyl 2-methyl 3-furyl methyl amine, n-methyl-1-2-methylfuran-3-yl methanamine
N-methyl-1-(2-methylfuran-3-yl)methanamine

Specifications

N-methyl(2-methyl-3-furyl)methylamine
94.5
95%
C7H11NO
MFCD09702377
IDODRBWAEHXURE-UHFFFAOYSA-N
N-methyl-1-(2-methylfuran-3-yl)methanamine
24229548
95%
916766-85-3
100.0
Amber Glass Bottle
1g
n-methyl 2-methyl-3-furyl methylamine, methyl 2-methylfuran-3-yl methyl amine, n-methyl-2-methylfur-3-yl methylamine, 2-methyl-3-methylamino methyl furan, 3-furanmethanamine,n,2-dimethyl, 3-furanmethanamine, n,2-dimethyl, methyl 2-methyl 3-furyl methyl amine, n-methyl-1-2-methylfuran-3-yl methanamine
CC1=C(C=CO1)CNC
125.171
125.17
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Harmful if inhaled.
Causes severe skin burns and eye damage.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
Documents

Documents