missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-[1-(6-methylpyrazin-2-yl)piperid-3-yl]methylamine, 97%, Thermo Scientific™

Chemical Identifiers

CAS 941716-84-3
Molecular Formula C12H20N4
Molecular Weight (g/mol) 220.32
MDL Number MFCD09879920
InChI Key CBRQUNBWUQBINE-UHFFFAOYSA-N
Synonym n-methyl-1-6-methylpyrazin-2-yl piperid-3-yl methylamine, methyl 1-6-methylpyrazin-2-yl piperidin-3-yl methyl amine, methyl 1-6-methylpyrazin-2-yl 3-piperidyl methyl amine, n-methyl-1-1-6-methylpyrazin-2-yl piperidin-3-yl methanamine
PubChem CID 24229603
IUPAC Name N-methyl-1-[1-(6-methylpyrazin-2-yl)piperidin-3-yl]methanamine
SMILES CC1=CN=CC(=N1)N2CCCC(C2)CNC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
CC59046CB
View Documents Promotion Details
N/A 250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC59046DA
View Documents Promotion Details
N/A 1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

941716-84-3
220.32
CBRQUNBWUQBINE-UHFFFAOYSA-N
24229603
CC1=CN=CC(=N1)N2CCCC(C2)CNC
C12H20N4
MFCD09879920
n-methyl-1-6-methylpyrazin-2-yl piperid-3-yl methylamine, methyl 1-6-methylpyrazin-2-yl piperidin-3-yl methyl amine, methyl 1-6-methylpyrazin-2-yl 3-piperidyl methyl amine, n-methyl-1-1-6-methylpyrazin-2-yl piperidin-3-yl methanamine
N-methyl-1-[1-(6-methylpyrazin-2-yl)piperidin-3-yl]methanamine

Specifications

941716-84-3
100.0
C12H20N4
n-methyl-1-6-methylpyrazin-2-yl piperid-3-yl methylamine, methyl 1-6-methylpyrazin-2-yl piperidin-3-yl methyl amine, methyl 1-6-methylpyrazin-2-yl 3-piperidyl methyl amine, n-methyl-1-1-6-methylpyrazin-2-yl piperidin-3-yl methanamine
CC1=CN=CC(=N1)N2CCCC(C2)CNC
220.32
220.32
N-Methyl-[1-(6-methylpyrazin-2-yl)piperid-3-yl]methylamine
96.5
97%
MFCD09879920
CBRQUNBWUQBINE-UHFFFAOYSA-N
N-methyl-1-[1-(6-methylpyrazin-2-yl)piperidin-3-yl]methanamine
24229603
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
Documents

Documents