accessibility menu, dialog, popup

UserName

CAS RN 941716-84-3

CH3NNNNHH3C

CAS RN 941716-84-3

IUPAC Name: methyl({[1-(6-methylpyrazin-2-yl)piperidin-3-yl]methyl})amine
Synonyms: methyl({[1-(6-methylpyrazin-2-yl)piperidin-3-yl]methyl})amine
Molecular Weight (g/mol): 220.32
Molecular Formula: C12H20N4
InChi Key: CBRQUNBWUQBINE-UHFFFAOYNA-N
SMILES: CNCC1CCCN(C1)C1=NC(C)=CN=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)