missing translation for 'onlineSavingsMsg'
Learn More

N-Methoxydiacetamide 95.0+%, TCI America™

Catalog No. M11685G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M11685G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M11685G Supplier TCI America Supplier No. M11685G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 128459-09-6
Molecular Formula C5H9NO3
Molecular Weight (g/mol) 131.131
MDL Number MFCD00191669
InChI Key HWZKQIRJZPOEQF-UHFFFAOYSA-N
Synonym N,N,N-Diacetylmethoxyamine
PubChem CID 538412
IUPAC Name N-acetyl-N-methoxyacetamide
SMILES CC(=O)N(C(=O)C)OC

Specifications

Color White-Yellow
Boiling Point 74°C
Quantity 5 g
Formula Weight 131.13
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material N-Methoxydiacetamide [Selective Acetylating Reagent]