accessibility menu, dialog, popup

UserName

CH3ONOCH3OH3C

CAS RN 128459-09-6

IUPAC Name: N-acetyl-N-methoxyacetamide
Synonyms: N-acetyl-N-methoxyacetamide
Molecular Weight (g/mol): 131.13
Molecular Formula: C5H9NO3
InChi Key: HWZKQIRJZPOEQF-UHFFFAOYSA-N
SMILES: CON(C(C)=O)C(C)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results