missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Methoxycarbonylmaleimide 98.0+%, TCI America™
$139.81 - $488.38
Chemical Identifiers
| CAS | 55750-48-6 |
|---|---|
| Molecular Formula | C6H5NO4 |
| Molecular Weight (g/mol) | 155.109 |
| MDL Number | MFCD00042755 |
| InChI Key | LLAZQXZGAVBLRX-UHFFFAOYSA-N |
| PubChem CID | 580610 |
| IUPAC Name | methyl 2,5-dioxopyrrole-1-carboxylate |
| SMILES | COC(=O)N1C(=O)C=CC1=O |
Chemical Identifiers
| 55750-48-6 | |
| 155.109 | |
| LLAZQXZGAVBLRX-UHFFFAOYSA-N | |
| methyl 2,5-dioxopyrrole-1-carboxylate |
| C6H5NO4 | |
| MFCD00042755 | |
| 580610 | |
| COC(=O)N1C(=O)C=CC1=O |
Specifications
| 66°C | |
| C6H5NO4 | |
| LLAZQXZGAVBLRX-UHFFFAOYSA-N | |
| methyl 2,5-dioxopyrrole-1-carboxylate | |
| 155.109 | |
| 155.11 | |
| Crystalline Powder |
| 55750-48-6 | |
| MFCD00042755 | |
| COC(=O)N1C(=O)C=CC1=O | |
| 1 g | |
| 580610 | |
| ≥98.0% (N) | |
| N-Methoxycarbonylmaleimide |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator