accessibility menu, dialog, popup

UserName

CAS RN 55750-48-6

CH3OONOO

CAS RN 55750-48-6

IUPAC Name: methyl 2,5-dioxo-2,5-dihydro-1H-pyrrole-1-carboxylate
Synonyms: methyl 2,5-dioxopyrrole-1-carboxylate
Molecular Weight (g/mol): 155.11
Molecular Formula: C6H5NO4
InChi Key: LLAZQXZGAVBLRX-UHFFFAOYSA-N
SMILES: COC(=O)N1C(=O)C=CC1=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results