missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(Methoxycarbonyl)-L-valine 98.0+%, TCI America™
$67.38 - $115.51
Chemical Identifiers
| CAS | 74761-42-5 |
|---|---|
| Molecular Formula | C7H13NO4 |
| Molecular Weight (g/mol) | 175.184 |
| MDL Number | MFCD12795357 |
| InChI Key | CEFVHPDFGLDQKU-YFKPBYRVSA-N |
| PubChem CID | 10942966 |
| IUPAC Name | (2S)-2-(methoxycarbonylamino)-3-methylbutanoic acid |
| SMILES | CC(C)C(C(=O)O)NC(=O)OC |
Chemical Identifiers
| 74761-42-5 | |
| 175.184 | |
| CEFVHPDFGLDQKU-YFKPBYRVSA-N | |
| (2S)-2-(methoxycarbonylamino)-3-methylbutanoic acid |
| C7H13NO4 | |
| MFCD12795357 | |
| 10942966 | |
| CC(C)C(C(=O)O)NC(=O)OC |
Specifications
| 74761-42-5 | |
| White | |
| MFCD12795357 | |
| CEFVHPDFGLDQKU-YFKPBYRVSA-N | |
| (2S)-2-(methoxycarbonylamino)-3-methylbutanoic acid | |
| 10942966 | |
| ≥98.0% (HPLC,T) | |
| N-(Methoxycarbonyl)-L-valine |
| 111°C | |
| C7H13NO4 | |
| 5 g | |
| CC(C)C(C(=O)O)NC(=O)OC | |
| 175.184 | |
| 175.18 | |
| Crystalline Powder |
Safety and Handling
TSCA : No