missing translation for 'onlineSavingsMsg'
Learn More

N-Methoxy-N-methylpropionamide 97.0+%, TCI America™

Catalog No. m30961g
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M30961G 1 g
M30965G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M30961G Supplier TCI America Supplier No. M30961G
Only null left

Chemical Identifiers

CAS 104863-65-2
Molecular Formula C5H11NO2
Molecular Weight (g/mol) 117.15
MDL Number MFCD09833541
InChI Key YKVJZSZZQKQJMO-UHFFFAOYSA-N
Synonym N-Methoxy-N-methylpropanamide
PubChem CID 11344007
IUPAC Name N-methoxy-N-methylpropanamide
SMILES CCC(=O)N(C)OC

Specifications

Color Colorless
Quantity 1 g
UN Number 1993
Formula Weight 117.15
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material N-Methoxy-N-methylpropionamide