accessibility menu, dialog, popup

UserName

H3CONCH3OCH3

CAS RN 104863-65-2

IUPAC Name: N-methoxy-N-methylpropanamide
Synonyms: N-methoxy-N-methylpropanamide
Molecular Weight (g/mol): 117.15
Molecular Formula: C5H11NO2
InChi Key: YKVJZSZZQKQJMO-UHFFFAOYSA-N
SMILES: CCC(=O)N(C)OC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results