missing translation for 'onlineSavingsMsg'
Learn More

N-Methoxy-N-methylbenzamide 98.0+%, TCI America™

$143.54 - $471.60

Chemical Identifiers

CAS 6919-61-5
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.192
MDL Number MFCD00075320
InChI Key UKERDACREYXSIV-UHFFFAOYSA-N
Synonym n-methyl-n-methoxybenzamide, benzamide, n-methoxy-n-methyl, n-methoxy-n-methyl-benzamide, methyl n-methylbenzohydroxamate, pubchem17496, acmc-1b5bp, n-methoxy-n-methylbenzamid, n-methoxy-n-methyl benzamide, ksc445c3n, n-methoxy-n-methylbenzamide
PubChem CID 569575
ChEBI CHEBI:59742
IUPAC Name N-methoxy-N-methylbenzamide
SMILES CN(C(=O)C1=CC=CC=C1)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M26725G
View Documents
TCI America
M26725G
5 g N/A
Only null left
 
M267225G
View Documents
TCI America
M267225G
25 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

6919-61-5
165.192
UKERDACREYXSIV-UHFFFAOYSA-N
569575
N-methoxy-N-methylbenzamide
C9H11NO2
MFCD00075320
n-methyl-n-methoxybenzamide, benzamide, n-methoxy-n-methyl, n-methoxy-n-methyl-benzamide, methyl n-methylbenzohydroxamate, pubchem17496, acmc-1b5bp, n-methoxy-n-methylbenzamid, n-methoxy-n-methyl benzamide, ksc445c3n, n-methoxy-n-methylbenzamide
CHEBI:59742
CN(C(=O)C1=CC=CC=C1)OC

Specifications

6919-61-5
112°C
MFCD00075320
n-methyl-n-methoxybenzamide, benzamide, n-methoxy-n-methyl, n-methoxy-n-methyl-benzamide, methyl n-methylbenzohydroxamate, pubchem17496, acmc-1b5bp, n-methoxy-n-methylbenzamid, n-methoxy-n-methyl benzamide, ksc445c3n, n-methoxy-n-methylbenzamide
CN(C(=O)C1=CC=CC=C1)OC
165.192
CHEBI:59742
≥98.0% (GC)
N-Methoxy-N-methylbenzamide
Colorless
C9H11NO2
5 g
UKERDACREYXSIV-UHFFFAOYSA-N
N-methoxy-N-methylbenzamide
569575
165.19
Liquid
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'