missing translation for 'onlineSavingsMsg'
Learn More

N-(m-Toluoyl)glycine 98.0+%, TCI America™

Catalog No. T03125G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T03125G 5 g
T031225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T03125G Supplier TCI America Supplier No. T03125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 27115-49-7
Molecular Formula C10H11NO3
Molecular Weight (g/mol) 193.20
MDL Number MFCD00044399
InChI Key YKAKNMHEIJUKEX-UHFFFAOYSA-N
Synonym 3-methylhippuric acid, m-methylhippuric acid, n-m-toluoyl glycine, m-toluric acid, n-3-methylbenzoyl glycine, 3-methylbenzoylglycine, glycine, n-3-methylbenzoyl, meta-methylhippuric acid, hippuric acid, m-methyl, 3-methyl hippuric acid
PubChem CID 99223
ChEBI CHEBI:68500
IUPAC Name 2-[(3-methylphenyl)formamido]acetic acid
SMILES CC1=CC=CC(=C1)C(=O)NCC(O)=O

Specifications

Melting Point 138°C
Color White
Quantity 5 g
Formula Weight 193.20
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-(m-Toluoyl)glycine