missing translation for 'onlineSavingsMsg'
Learn More

N-Lauryldiethanolamine 95.0+%, TCI America™

Catalog No. L00205G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L00205G 5 g
L002025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. L00205G Supplier TCI America Supplier No. L00205G
Only null left

Chemical Identifiers

CAS 1541-67-9
Molecular Formula C16H34NO2
Molecular Weight (g/mol) 272.45
MDL Number MFCD00059002
InChI Key ASGZLXKBMOBPBV-UHFFFAOYSA-N
PubChem CID 352309
IUPAC Name dodecylidenebis(2-hydroxyethyl)azanium
SMILES CCCCCCCCCCCC=[N+](CCO)CCO

Specifications

Boiling Point 195°C
Quantity 5 g
Synonym N,N-Bis(2-hydroxyethyl)dodecylamine, N-Dodecyldiethanolamine, 2,2′C-(Dodecylazanediyl)diethanol
Formula Weight 273.46
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material N-Lauryldiethanolamine
EINECSNumber (2)-0353&(7)-0060
TSCA Yes