CAS RN 1541-67-9
IUPAC Name:
dodecylidenebis(2-hydroxyethyl)azanium
Synonyms:
N-lauryl diethanolamine
Molecular Weight (g/mol):
272.45
Molecular Formula:
C16H34NO2
InChi Key:
ASGZLXKBMOBPBV-UHFFFAOYSA-N
SMILES:
CCCCCCCCCCCC=[N+](CCO)CCO
1
–
1
of
1
results
1
N-Lauryldiethanolamine 95.0+%, TCI America™
CAS: 1541-67-9 Molecular Formula: C16H34NO2 Molecular Weight (g/mol): 272.45 MDL Number: MFCD00059002 InChI Key: ASGZLXKBMOBPBV-UHFFFAOYSA-N PubChem CID: 352309 IUPAC Name: dodecylidenebis(2-hydroxyethyl)azanium SMILES: CCCCCCCCCCCC=[N+](CCO)CCO