missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N'-Isopropylidene-2-nitrobenzenesulfonohydrazide 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 136°C |
| Quantity | 5 g |
| Formula Weight | 257.26 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | N′-Isopropylidene-2-nitrobenzenesulfonohydrazide |
Safety and Handling
| TSCA | No |
| Recommended Storage | Freezer |